Merck
  • Inhibition of MAPK/ERK pathway promotes oligodendrocytes generation and recovery of demyelinating diseases.
Inhibition of MAPK/ERK pathway promotes oligodendrocytes generation and recovery of demyelinating diseases.
Glia (2019-03-01)
Na Suo, Yu-E Guo, Bingqing He, Haifeng Gu, Xin Xie
ABSTRACT
Oligodendrocytes (OLs) are the myelinating glia of the central nervous system. Injury to OLs causes myelin loss. In demyelinating diseases, such as multiple sclerosis, the remyelination is hindered principally due to a failure of the oligodendrocyte precursor cells (OPCs) to differentiate into mature OLs. To identify inducers of OPC to OL differentiation, a high-throughput screening based on myelin basic protein expression using neural progenitor cells-derived OPCs has been performed and, PD0325901-an MEK (MAPK kinase) inhibitor-is found to significantly enhance OPC to OL differentiation in a dose- and time-dependent manner. Other MEK inhibitors also display similar effect, indicating blockade of MAPK-ERK signaling is sufficient to induce OPC differentiation into OLs. PD0325901 facilitates the formation of myelin sheaths in OPC-neuron co-culture in vitro. And in experimental autoimmune encephalomyelitis model and cuprizone-induced demyelination model, PD0325901 displays significant therapeutic effect by promoting myelin regeneration. Our results suggest that targeting the MAPK-ERK pathway might be an intriguing way to develop new therapies for demyelinating diseases.
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
ESGRO Complete Accutase, The ESGRO Complete Accutase is a cell detachment solution of proteolytic & collagenolytic enzymes, qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete Clonal Grade Medium.