Merck
  • The D2-like Dopamine Receptor Agonist Quinpirole Microinjected Into the Ventral Pallidum Dose-Dependently Inhibits the VTA and Induces Place Aversion.
The D2-like Dopamine Receptor Agonist Quinpirole Microinjected Into the Ventral Pallidum Dose-Dependently Inhibits the VTA and Induces Place Aversion.
The international journal of neuropsychopharmacology (2022-03-30)
Laszlo Peczely, Tamas Ollmann, Kristof Laszlo, Laszlo Lenard, Anthony A Grace
ABSTRACT
The ventral pallidum (VP) is a dopaminoceptive forebrain structure regulating the ventral tegmental area (VTA) dopaminergic population activity. We have recently demonstrated that in the VP, the D2-like dopamine (DA) receptor agonist quinpirole dose dependently facilitates memory consolidation in inhibitory avoidance and spatial learning. According to our hypothesis, quinpirole microinjected into the VP can modulate the VTA DAergic activity and influence motivation and learning processes of rats. Quinpirole was microinjected at 3 different doses into the VP of male rats, and controls received vehicle. Single unit recordings were employed to assess VTA DAergic activity. To investigate the possible reinforcing or aversive effect of quinpirole in the VP, the conditioned place preference paradigm was used. Our results showed that intra-VP quinpirole microinjection regulates VTA DAergic neurons according to an inverted U-shaped dose-response curve. The largest dose of quinpirole decreased the population activity and strongly reduced burst activity of the DAergic neurons in the first hour after its application. In contrast, the 2 smaller doses increased DA population activity, but their effect started with a delay 1 hour after their microinjection. The CPP experiments revealed that the largest dose of quinpirole in the VP induced place aversion in the rats. Furthermore, the largest dose of quinpirole induced an acute locomotor activity reduction, while the medium dose led to a long-duration increase in locomotion. In summary, quinpirole dose dependently regulates VTA DAergic activity as well as the motivation and motor behavior of the rats at the level of the VP.
Sigma-Aldrich
(−)-Quinpirole hydrochloride, ≥98% (HPLC), solid