- Simultaneous reversed-phase high-performance liquid chromatographic separation of mono-, di-and trichloroanilines through a gradient elution optimised by experimental design.
Simultaneous reversed-phase high-performance liquid chromatographic separation of mono-, di-and trichloroanilines through a gradient elution optimised by experimental design.
Journal of chromatography. A (2002-02-28)
M C Gennaro, E Marengo, V Gianotti, S Angioi
PMID11860144
ABSTRACT
A new RP-HPLC method for the simultaneous determination of the 13 mono-, di- and trichloroanilines has been developed. In order to obtain the analyte resolution within an acceptable analysis time, a gradient elution program has been optimised through the use of an experimental design and a grid search algorithm. The optimized conditions provided the resolution of all the analytes in less than 80 min. The primary validation of the analytical method gave limit of detection values ranging between 0.02 and 0.06 mg/l and very good linearity of the calibration curves.