Merck
  • Fluorescent Brighteners as Visible LED-Light Sensitive Photoinitiators for Free Radical Photopolymerizations.
Fluorescent Brighteners as Visible LED-Light Sensitive Photoinitiators for Free Radical Photopolymerizations.
Macromolecular rapid communications (2016-04-14)
Xiaoling Zuo, Fabrice Morlet-Savary, Bernadette Graff, Nicolas Blanchard, Jean-Philippe Goddard, Jacques Lalevée
ABSTRACT
The photochemical and electrochemical investigations of commercially available, safe, and cheap fluorescent brighteners, namely, triazinylstilbene (commercial name: fluorescent brightener 28) and 2,5-bis(5-tert-butyl-benzoxazol-2-yl)thiophene, as well as their original use as photoinitiators of polymerization upon light emitting diode (LED) irradiation are reported. Remarkably, their excellent near-UV-visible absorption properties combined with outstanding fluorescent properties allow them to act as high-performance photoinitiators when used in combination with diaryliodonium salt. These two-component photoinitiating systems can be employed for free radical polymerizations of acrylate. In addition, this brightener-initiated photopolymerization is able to overcome oxygen inhibition even upon irradiation with low LED light intensity. The underlying photochemical mechanisms are investigated by electron-spin resonance-spin trapping, fluorescence, cyclic voltammetry, and steady-state photolysis techniques.
Sigma-Aldrich
2,5-Bis(5-tert-butyl-benzoxazol-2-yl)thiophene, suitable for scintillation, 99%