Merck
  • T cell migration potentiates HIV infection by enhancing viral fusion and integration.
T cell migration potentiates HIV infection by enhancing viral fusion and integration.
Cell reports (2022-02-24)
Paul Lopez, Oluwaseun Ajibola, Amelie Pagliuzza, Romaniya Zayats, Wan Hon Koh, Alon Herschhorn, Nicolas Chomont, Thomas T Murooka
ABSTRACT
T cells actively migrate along reticular networks within lymphoid organs in search for cognate antigen, but how these behaviors impact HIV entry and infection is unclear. Here, we show that migratory T cells in 3D collagen matrix display significantly enhanced infection and integration by cell-free R5-tropic lab adapted and transmitted/founder molecular HIV clones in the absence of exogenous cytokines or cationic polymers. Using two different collagen matrices that either support or restrict T cell migration, we observe high levels of HIV fusion in migratory T cells, whereas non-motile T cells display low viral entry and integration. Motile T cells were less sensitive to combination antiretroviral drugs and were able to freely migrate into regions with high HIV densities, resulting in high infection rates. Together, our studies indicate that the environmental context in which initial HIV-T cell encounters occur modulates HIV-1 entry and integration efficiencies.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Probenecid
Sigma-Aldrich
Latrunculin A, Latrunculia magnifica, Latrunculin A, CAS 76343-93-6, is a cell-permeable marine toxin that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (KD = 200 nM).