Merck
  • Reductive TCA cycle metabolism fuels glutamine- and glucose-stimulated insulin secretion.
Reductive TCA cycle metabolism fuels glutamine- and glucose-stimulated insulin secretion.
Cell metabolism (2020-12-16)
Guo-Fang Zhang, Mette V Jensen, Sarah M Gray, Kimberley El, You Wang, Danhong Lu, Thomas C Becker, Jonathan E Campbell, Christopher B Newgard
ABSTRACT
Metabolic fuels regulate insulin secretion by generating second messengers that drive insulin granule exocytosis, but the biochemical pathways involved are incompletely understood. Here we demonstrate that stimulation of rat insulinoma cells or primary rat islets with glucose or glutamine + 2-aminobicyclo-(2,2,1)-heptane-2-carboxylic acid (Gln + BCH) induces reductive, "counter-clockwise" tricarboxylic acid (TCA) cycle flux of glutamine to citrate. Molecular or pharmacologic suppression of isocitrate dehydrogenase-2 (IDH2), which catalyzes reductive carboxylation of 2-ketoglutarate to isocitrate, results in impairment of glucose- and Gln + BCH-stimulated reductive TCA cycle flux, lowering of NADPH levels, and inhibition of insulin secretion. Pharmacologic suppression of IDH2 also inhibits insulin secretion in living mice. Reductive TCA cycle flux has been proposed as a mechanism for generation of biomass in cancer cells. Here we demonstrate that reductive TCA cycle flux also produces stimulus-secretion coupling factors that regulate insulin secretion, including in non-dividing cells.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
INS-1 832/13 Rat Insulinoma Cell Line, INS-1 832/13 rat insulinoma cell line is a useful model for insulin secretion regulation and pancreatic islet beta-cell function studies.