Merck
  • Binding of endostatin to human ovarian cancer cells inhibits cell attachment.
Binding of endostatin to human ovarian cancer cells inhibits cell attachment.
International journal of cancer (2007-06-29)
Yumi Yokoyama, S Ramakrishnan
ABSTRACT
Endostatin, a C-terminal fragment of collagen type XVIII, is one of the well-characterized endogenous inhibitors of angiogenesis. Endostatin is known to bind integrin alpha(5)beta(1), which is upregulated on tumor endothelium. Most of the ovarian cancer cells express significant amounts of alpha(5)beta(1) integrin, which is important for ovarian cancer cells to attach to the peritoneal wall. Therefore we investigated whether endostatin could directly bind ovarian cancer cells and inhibit tumor cell attachment to extracellular matrix. Binding of endostatin to ovarian cancer cells was characterized by preincubation with function blocking antibodies to integrin subunits. These studies showed that ovarian cancer cell attachment to fibronectin-coated wells can be inhibited by alpha(5)beta(1) integrin specific antibodies as well as endostatin. Downregulation of integrin alpha(5) and beta(1) by siRNA abrogated the binding of OVCAR5 and human umbilical vein endothelial cell to endostatin. Although endostatin treatment did not affect ovarian cancer cell migration, treated cells failed to attach mouse peritoneal wall preparations. These studies suggest an extra-antiangiogenic role for endostatin, which can be used prevent peritoneal attachment and dissemination of ovarian cancer cells.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone 25E11, clone 25E11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α3β1 Antibody, clone M-KID2, clone M-KID2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone ASC-6, clone ASC-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2β1 Antibody, clone BHA2.1, clone BHA2.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, clone 6S6, Chemicon®, from mouse