Merck
  • Recombinant sucrose phosphorylase from Leuconostoc mesenteroides: characterization, kinetic studies of transglucosylation, and application of immobilised enzyme for production of alpha-D-glucose 1-phosphate.
Recombinant sucrose phosphorylase from Leuconostoc mesenteroides: characterization, kinetic studies of transglucosylation, and application of immobilised enzyme for production of alpha-D-glucose 1-phosphate.
Journal of biotechnology (2007-01-12)
Christiane Goedl, Alexandra Schwarz, Alphonse Minani, Bernd Nidetzky
ABSTRACT
Sucrose phosphorylase catalyzes the reversible conversion of sucrose (alpha-D-glucopyranosyl-1,2-beta-D-fructofuranoside) and phosphate into D-fructose and alpha-D-glucose 1-phosphate. We report on the molecular cloning and expression of the structural gene encoding sucrose phosphorylase from Leuconostoc mesenteroides (LmSPase) in Escherichia coli DH10B. The recombinant enzyme, containing an 11 amino acid-long N-terminal metal affinity fusion peptide, was overproduced 60-fold in comparison with the natural enzyme. It was purified to apparent homogeneity using copper-loaded Chelating Sepharose and obtained in 20% yield with a specific activity of 190 Umg(-1). LmSPase was covalently attached onto Eupergit C with a binding efficiency of 50% and used for the continuous production of alpha-D-glucose 1-phosphate from sucrose and phosphate (600 mM each) in a packed-bed immobilised enzyme reactor (30 degrees C, pH 7.0). The reactor was operated at a stable conversion of 91% (550 mM product) and productivity of approximately 11 gl(-1)h(-1) for up to 600 h. A kinetic study of transglucosylation by soluble LmSPase was performed using alpha-d-glucose 1-phosphate as the donor substrate and various alcohols as acceptors. D- and L-arabitol were found to be good glucosyl acceptors.
Sigma-Aldrich
Sucrose Phosphorylase, recombinant, expressed in E. coli, lyophilized powder, ≥45 units/mg solid