Merck
  • Aliphatic C-H activation with aluminium trichloride-acetyl chloride: expanding the scope of the Baddeley reaction for the functionalisation of saturated hydrocarbons.
Aliphatic C-H activation with aluminium trichloride-acetyl chloride: expanding the scope of the Baddeley reaction for the functionalisation of saturated hydrocarbons.
Organic & biomolecular chemistry (2012-12-25)
Catherine L Lyall, Mario Uosis-Martin, John P Lowe, Mary F Mahon, G Dan Pantoş, Simon E Lewis
ABSTRACT
The functionalisation of decalin by means of an "aliphatic Friedel-Crafts" reaction was reported over fifty years ago by Baddeley et al. This protocol is of current relevance in the context of C-H activation and here we demonstrate its applicability to a range of other saturated hydrocarbons. Structural elucidation of the products is described and a mechanistic rationale for their formation is presented. The "aliphatic Friedel-Crafts" procedure allows for production of novel oxygenated building blocks from abundant hydrocarbons and as such can be considered to add significant synthetic value in a single step.
Sigma-Aldrich
Bicyclohexyl, 99%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%