Merck
  • Interaction of Hepatitis B Virus X Protein with the Pregnane X Receptor Enhances the Synergistic Effects of Aflatoxin B1 and Hepatitis B Virus on Promoting Hepatocarcinogenesis.
Interaction of Hepatitis B Virus X Protein with the Pregnane X Receptor Enhances the Synergistic Effects of Aflatoxin B1 and Hepatitis B Virus on Promoting Hepatocarcinogenesis.
Journal of clinical and translational hepatology (2021-08-28)
Yongdong Niu, Shaohua Fan, Qin Luo, Liming Chen, Danmei Huang, Wenjun Chang, Wenxin Qin, Ganggang Shi
ABSTRACT
Hepatitis B virus (HBV) infection has been found to increase hepatocellular sensitivity to carcinogenic xenobiotics, by unknown mechanisms, in the generation of hepatocellular carcinoma. The pregnane X receptor (PXR) is a key regulator of the body's defense against xenobiotics, including xenobiotic carcinogens and clinical drugs. In this study, we aimed to investigate the molecular mechanisms of HBV X protein (HBx)-PXR signaling in the synergistic effects of chemical carcinogens in HBV-associated hepatocarcinogenesis. The expression profile of PXR-cytochrome p450 3A4 (CYP3A4) signaling was determined by PCR, western blotting, and tissue microarray. Cell viability and aflatoxin B1 (AFB1) cytotoxicity were measured using the cell counting kit-8 assay. Target gene expression was evaluated using transient transfection and real time-PCR. The genotoxicity of AFB1 was assessed in newborn mice with a single dose of AFB1. HBx enhanced the hepatotoxicity of AFB1 by activating CYP3A4 and reducing glutathione S-transferase Mu 1 (GSTM1) in cell lines. Activation of PXR by pregnenolone 16α-carbonitrile increased AFB1-induced liver tumor incidence by up-regulating oncogenic KRAS to enhance interleukin (IL)-11:IL-11 receptor subunit alpha-1 (IL11RA-1)-mediated inflammation in an HBx transgenic model. Our finding regarding AFB1 toxicity enhancement by an HBx-PXR-CYP3A4/ GSTM1-KRAS-IL11:IL11RA signaling axis provides a rational explanation for the synergistic effects of chemical carcinogens in HBV infection-associated hepatocarcinogenesis.
Sigma-Aldrich
Anti-Hepatitis B Virus Antibody, X-Protein, a.a. 90-115, clone 227, clone 227, Chemicon®, from mouse