Merck
  • Synthesis of the bis-cystinyl-fragment 225-232/225'-232' of the human IgGl hinge region.
Synthesis of the bis-cystinyl-fragment 225-232/225'-232' of the human IgGl hinge region.
International journal of peptide and protein research (1988-11-01)
E Wünsch, L Moroder, S Göhring-Romani, H J Musiol, W Göhring, G Bovermann
ABSTRACT
In human IgGl the two heavy chains are crosslinked in the central portion of the molecule by two disulfide bridges forming a double chain bis-cystinyl cyclic peptide in parallel alignment. For our synthetic studies we have chosen the sequence portion 225-232/225'-232', i.e. [H-Thr-C1ys-Pro-Pro-C1ys-Pro-Ala-Pro-OH]2. By the use of a combination of the S-tert.-butylthio and the S-acetamidomethyl groups selective cysteine pairings in two successive steps produced the hinge hexadecapeptide in parallel and antiparallel alignment as homogeneous and well characterized compounds. Thiol disulfide interchange experiments on the antiparallel dimer led to over 90% conversion to the parallel isomer. Similarly random air-oxidation was found to generate again mainly the parallel dimer, thus strongly suggesting that this sequence portion contains sufficient structural information for a correct assembly of the two heavy chains of immunoglobulins without decisive contribution of a protein disulfide isomerase.
Sigma-Aldrich
S-tert-Butylmercapto-L-cysteine