Merck
  • Structural characterisation of the HT3 motif of the polyhistidine triad protein D from Streptococcus pneumoniae.
Structural characterisation of the HT3 motif of the polyhistidine triad protein D from Streptococcus pneumoniae.
FEBS letters (2018-06-02)
Zhenyao Luo, Victoria G Pederick, James C Paton, Christopher A McDevitt, Bostjan Kobe
ABSTRACT
The bacterium Streptococcus pneumoniae (the pneumococcus) is a major human pathogen that requires Zn2+ for its survival and virulence in the host environment. Polyhistidine triad protein D (PhtD) has a known role in pneumococcal Zn2+ homeostasis. However, the mechanistic basis of PhtD function remains unclear, partly due to a lack of structural information. Here, we determined the crystal structure of the fragment PhtD269-339 , containing the third Zn2+ -binding histidine triad (HT) motif of the protein. Analysis of the structure suggests that Zn2+ binding occurs at the surface of the protein and that all five HT motifs in the protein bind Zn2+ and share similar structures. These new structural insights aid in our understanding of how the Pht proteins facilitate pneumococcal Zn2+ acquisition.
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein