Merck
  • Structure-activity relationship of salicylic acid derivatives on inhibition of TNF-α dependent NFκB activity: Implication on anti-inflammatory effect of N-(5-chlorosalicyloyl)phenethylamine against experimental colitis.
Structure-activity relationship of salicylic acid derivatives on inhibition of TNF-α dependent NFκB activity: Implication on anti-inflammatory effect of N-(5-chlorosalicyloyl)phenethylamine against experimental colitis.
European journal of medicinal chemistry (2011-12-14)
Jihye Kim, Sookjin Kang, Sungchae Hong, Soowhan Yum, Young Mi Kim, Yunjin Jung
ABSTRACT
To develop a more potent NFκB inhibitor from salicylic acid which is known to inhibit activity of NFκB, a transcription factor regulating genes involved in immunity, inflammation and tumorigenesis, derivatives of salicylic acid (SA) where the 5 position, carboxyl or hydroxyl group was modified were treated in HCT116 cells transfected with an NFκB dependent luciferase gene and LPS-stimulated RAW264.7 cells. Amidation of the carboxylic group or substitution of chlorine at the 5 position increased the ability of SA to suppress the expression of NFκB dependent luciferase and inducible nitric oxide synthase, a product of an NFκB target gene. Moreover, simultaneous amidation and chlorination of SA (5-chlorosalicylamide; 5-CSAM) conferred an additive NFκB inhibitory activity on SA. To further enhance the inhibitory activity, N-modification was imposed on 5-CSAM. N-(5-chlorosalicyloyl)phenethylamine (5-CSPA), N-(5-chlorosalicyloyl)3-phenylpropylamine (5-CSPPA) and N-(5-chlorosalicyloyl)4-hydroxyphenylethylamine (5-CSHPA) showed greater potencies for inhibiting NFκB activity than other derivatives. Their IC(50)s' in the luciferase assay measured 15μM (5-CSPA), 17μM (5-CSPPA) and 91μM (5-CSHPA). Rectal administration of 5-CSPA ameliorated TNBS-induced rat colitis, which was more effective than a conventional drug, 5-aminosalicylic acid. These data may provide useful information for development of a therapeutic agent for treatment of diseases where NFκB plays a critical role in the pathogenic progresses.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylamide, 99%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.
Supelco
Methyl salicylate, analytical standard
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Chlorosalicylic acid, 98%
Sigma-Aldrich
Aspirin, meets USP testing specifications