Merck
  • Parp1 hyperactivity couples DNA breaks to aberrant neuronal calcium signalling and lethal seizures.
Parp1 hyperactivity couples DNA breaks to aberrant neuronal calcium signalling and lethal seizures.
EMBO reports (2021-05-02)
Emilia Komulainen, Jack Badman, Stephanie Rey, Stuart Rulten, Limei Ju, Kate Fennell, Ilona Kalasova, Kristyna Ilievova, Peter J McKinnon, Hana Hanzlikova, Kevin Staras, Keith W Caldecott
ABSTRACT
Defects in DNA single-strand break repair (SSBR) are linked with neurological dysfunction but the underlying mechanisms remain poorly understood. Here, we show that hyperactivity of the DNA strand break sensor protein Parp1 in mice in which the central SSBR protein Xrcc1 is conditionally deleted (Xrcc1Nes-Cre ) results in lethal seizures and shortened lifespan. Using electrophysiological recording and synaptic imaging approaches, we demonstrate that aberrant Parp1 activation triggers seizure-like activity in Xrcc1-defective hippocampus ex vivo and deregulated presynaptic calcium signalling in isolated hippocampal neurons in vitro. Moreover, we show that these defects are prevented by Parp1 inhibition or deletion and, in the case of Parp1 deletion, that the lifespan of Xrcc1Nes-Cre mice is greatly extended. This is the first demonstration that lethal seizures can be triggered by aberrant Parp1 activity at unrepaired SSBs, highlighting PARP inhibition as a possible therapeutic approach in hereditary neurological disease.
Product Description
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-poly-ADP-ribose binding reagent, Anti-poly-ADP-ribose binding reagent is a reagent that selectively binds to ADP ribose for use in Western Blotting, Immunocytochemistry and Dot Blot.
Sigma-Aldrich
Anti-pan-ADP-ribose binding reagent, from Escherichia coli