Merck
  • Quantification of systemic o-toluidine after intrathecal administration of hyperbaric prilocaine in humans: a prospective cohort study.
Quantification of systemic o-toluidine after intrathecal administration of hyperbaric prilocaine in humans: a prospective cohort study.
Archives of toxicology (2021-01-22)
Emmanuel Guntz, Andrea Carini, Stephan Koslitz, Thomas Brüning, Panayota Kapessidou, Tobias Weiss
ABSTRACT
Hyperbaric 2% prilocaine is increasingly used for spinal anesthesia. It is the only local anesthetic metabolized to o-toluidine, a human bladder carcinogen. Increase of o-toluidine hemoglobin adducts, a marker of o-toluidine ability to modify the DNA structure, was described following subcutaneous injection. In this prospective cohort study we aimed to assess and quantify o-toluidine hemoglobin adducts and urinary o-toluidine after a single intrathecal dose of hyperbaric prilocaine.10 patients undergoing surgery received 50 mg of hyperbaric prilocaine intrathecally. Blood and urine samples were collected before injection and up to 24 h later (Hospital Braine l'Alleud-Waterloo, Braine l'Alleud, Belgium). Urinary o-toluidine and o-toluidine hemoglobin adducts were measured by tandem mass-spectrometry after gas-chromatographic separation (Institute of the Ruhr-Universität, Bochum Germany). The trial was registered to ClinicalTrials.gov (NCT03642301; 22-08-2018)Intrathecal administration of 50 mg of hyperbaric prilocaine leads to a significant increase of o-toluidine hemoglobin adducts (0.1 ± 0.02-11.9 ± 1.9 ng/g Hb after 24 h, p = 0.001). Peak of urinary o-toluidine was observed after 8 h (0.1 ± 0.1-460.5 ± 352.8 µg/L, p = 0.001) and declined to 98 ± 66.8 µg/L after 24 h (mean ± SD)Single intrathecal administration of hyperbaric prilocaine leads to a systemic burden with o-toluidine and o-toluidine hemoglobin adducts. O-toluidine-induced modifications of DNA should be examined and intrathecal hyperbaric prilocaine should not be proposed to patients chronically exposed to o-toluidine.Clinical trial number and registry URL NCT03642301.