Merck
  • Structure-activity relationship studies of acridones as potential antipsoriatic agents. 1. Synthesis and antiproliferative activity of simple N-unsubstituted 10H-acridin-9-ones against human keratinocyte growth.
Structure-activity relationship studies of acridones as potential antipsoriatic agents. 1. Synthesis and antiproliferative activity of simple N-unsubstituted 10H-acridin-9-ones against human keratinocyte growth.
European journal of medicinal chemistry (2010-05-11)
Aleksandar Putic, Lambert Stecher, Helge Prinz, Klaus Müller
ABSTRACT
A series of N-unsubstituted hydroxy-10H-acridin-9-ones were synthesized and evaluated for inhibitory action against HaCaT keratinocyte growth, in order to explore their potential as antipsoriatic agents. For structure-activity relationship studies, the number and position of the hydroxyl groups were modified, the oxygen functions substituted or replaced, or additional functional groups were introduced into the acridone scaffold. 1,8-Dihydroxy-10H-acridin-9-one (4), which is an aza-analogue of the antipsoriatic anthralin, was only marginally active. However, 1,3-dihydroxy-substituted 5ee was the most potent acridone within this series and inhibited keratinocyte growth with an IC(50) value comparable to that of anthralin. In contrast to anthralin, nearly all members of the acridone series were devoid of radical generating properties, which were determined by their capability to interact with the free radical 2,2-diphenyl-1-picrylhydrazyl. Structures with a phenolic hydroxyl or an aromatic amine arranged ortho or para to the acridone NH group were exceptions. Also in contrast to anthralin, membrane-damaging effects as documented by the release of lactate dehydrogenase into the culture medium were not observed for acridones.
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Dithranol, ≥90% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
9(10H)-Acridanone, 99%
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)