Merck
Dendritic Localization and Exocytosis of NAAG in the Rat Hippocampus.
Cerebral cortex (New York, N.Y. : 1991) (2019-09-11)
K Nordengen, C Morland, B S Slusher, V Gundersen
ABSTRACT
While a lot is known about classical, anterograde neurotransmission, less is known about the mechanisms and molecules involved in retrograde neurotransmission. Our hypothesis is that N-acetylaspartylglutamate (NAAG), the most abundant dipeptide in the brain, may act as a retrograde transmitter in the brain. NAAG was predominantly localized in dendritic compartments of glutamatergic synapses in the intact hippocampus, where it was present in close proximity to synaptic-like vesicles. In acute hippocampal slices, NAAG was depleted from postsynaptic dendritic elements during neuronal stimulation induced by depolarizing concentrations of potassium or by exposure to glutamate receptor (GluR) agonists. The depletion was completely blocked by botulinum toxin B and strictly dependent on extracellular calcium, indicating exocytotic release. In contrast, there were low levels of NAAG and no effect by depolarization or GluR agonists in presynaptic glutamatergic terminals or GABAergic pre- and postsynaptic elements. Together these data suggest a possible role for NAAG as a retrograde signaling molecule at glutamatergic synapses via exocytotic release.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
β-Alanine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
N-Acetyl-Asp-Glu, ≥97% (TLC)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%