Merck
  • CD34+CD146+ adipose-derived stromal cells enhance engraftment of transplanted fat.
CD34+CD146+ adipose-derived stromal cells enhance engraftment of transplanted fat.
Stem cells translational medicine (2020-06-17)
Mimi R Borrelli, Ronak A Patel, Charles Blackshear, Stephanie Vistnes, Nestor M Diaz Deleon, Sandeep Adem, Abra H Shen, Jan Sokol, Arash Momeni, Dung Nguyen, Michael T Longaker, Derrick C Wan
ABSTRACT
Fat grafting is a surgical technique able to reconstruct and regenerate soft tissue. The adipose-derived stromal cells (ASCs) within the stromal vascular fraction are believed to drive these beneficial effects. ASCs are increasingly recognized to be a heterogeneous group, comprised of multiple stem and progenitor subpopulations with distinct functions. We hypothesized the existence of an ASC subpopulation with enhanced angiogenic potential. Human ASCs that were CD34+CD146+, CD34+CD146-, or CD34+ unfractionated (UF) were isolated by flow cytometry for comparison of expression of proangiogenic factors and endothelial tube-forming potential. Next, lipoaspirate was enriched with either CD34+CD146+, CD34+CD146-, CD34+ UF ASCs, or was not enriched, and grafted beneath the scalp skin of immunodeficient CD-1 Nude mice (10 000 cells/200 μL/graft). Fat retention was monitored radiographically more than 8 weeks and fat grafts were harvested for histological assessment of quality and vascularization. The CD34+CD146+ subpopulation comprised ~30% of ASCs, and exhibited increased expression of vascular endothelial growth factor and angiopoietin-1 compared to CD34+CD146- and CD34+ UF ASCs, and increased expression of fibroblast growth factor-2 compared to CD34+CD146- ASCs. The CD34+CD146+ subpopulation exhibited enhanced induction of tube-formation compared to CD34+CD146- ASCs. Upon transplantation, fat enriched CD34+CD146+ ASCs underwent less resorption and had improved histologic quality and vascularization. We have identified a subpopulation of CD34+ ASCs with enhanced angiogenic effects in vitro and in vivo, likely mediated by increased expression of potent proangiogenic factors. These findings suggest that enriching lipoaspirate with CD34+CD146+ ASCs may enhance fat graft vascularization and retention in the clinical setting.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Medium 199, Modified, with Earle′s salts, without L-glutamine, sodium bicarbonate, and phenol red, powder, suitable for cell culture