Merck
  • Identification, quantification and bioinformatic analysis of RNA-dependent proteins by RNase treatment and density gradient ultracentrifugation using R-DeeP.
Identification, quantification and bioinformatic analysis of RNA-dependent proteins by RNase treatment and density gradient ultracentrifugation using R-DeeP.
Nature protocols (2020-02-26)
Maiwen Caudron-Herger, Elsa Wassmer, Isha Nasa, Astrid-Solveig Schultz, Jeanette Seiler, Arminja N Kettenbach, Sven Diederichs
ABSTRACT
Analysis of RNA-protein complexes is central to understanding the molecular circuitry governing cellular processes. In recent years, several proteome-wide studies have been dedicated to the identification of RNA-binding proteins. Here, we describe in detail R-DeeP, an approach built on RNA dependence, defined as the ability of a protein to engage in protein complexes only in the presence of RNA, involving direct or indirect interaction with RNA. This approach provides-for the first time, to our knowledge-quantitative information on the fraction of a protein associated with RNA-protein complexes. R-DeeP is independent of any potentially biased purification procedures. It is based on cellular lysate fractionation by density gradient ultracentrifugation and subsequent analysis by proteome-wide mass spectrometry (MS) or individual western blotting. The comparison of lysates with and without previous RNase treatment enables the identification of differences in the apparent molecular weight and, hence, the size of the complexes. In combination with information from databases of protein-protein complexes, R-DeeP facilitates the computational reconstruction of protein complexes from proteins migrating in the same fraction. In addition, we developed a pipeline for the statistical analysis of the MS dataset to automatically identify RNA-dependent proteins (proteins whose interactome depends on RNA). With this protocol, the individual analysis of proteins of interest by western blotting can be completed within 1-2 weeks. For proteome-wide studies, additional time is needed for the integration of the proteomic and statistical analyses. In the future, R-DeeP can be extended to other fractionation techniques, such as chromatography.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Supelco
Bicinchoninic Acid solution
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%