MilliporeSigma
  • Targeted histone demethylation improves somatic cell reprogramming into cloned blastocysts but not postimplantation bovine concepti†.
Targeted histone demethylation improves somatic cell reprogramming into cloned blastocysts but not postimplantation bovine concepti†.
Biology of reproduction (2020-04-23)
Fanli Meng, Kathrin Stamms, Romina Bennewitz, Andria Green, Fleur Oback, Pavla Turner, Jingwei Wei, Björn Oback
ABSTRACT
Correct reprogramming of epigenetic marks in the donor nucleus is a prerequisite for successful cloning by somatic cell transfer (SCT). In several mammalian species, repressive histone (H) lysine (K) trimethylation (me3) marks, in particular H3K9me3, form a major barrier to somatic cell reprogramming into pluripotency and totipotency. We engineered bovine embryonic fibroblasts (BEFs) for the doxycycline-inducible expression of a biologically active, truncated form of murine Kdm4b, a demethylase that removes H3K9me3 and H3K36me3 marks. Upon inducing Kdm4b, H3K9me3 and H3K36me3 levels were reduced about 3-fold and 5-fold, respectively, compared with noninduced controls. Donor cell quiescence has been previously associated with reduced somatic trimethylation levels and increased cloning efficiency in cattle. Simultaneously inducing Kdm4b expression (via doxycycline) and quiescence (via serum starvation) further reduced global H3K9me3 and H3K36me3 levels by a total of 18-fold and 35-fold, respectively, compared with noninduced, nonstarved control fibroblasts. Following SCT, Kdm4b-BEFs reprogrammed significantly better into cloned blastocysts than noninduced donor cells. However, detrimethylated donors and sustained Kdm4b-induction during embryo culture did not increase the rates of postblastocyst development from implantation to survival into adulthood. In summary, overexpressing Kdm4b in donor cells only improved their reprogramming into early preimplantation stages, highlighting the need for alternative experimental approaches to reliably improve somatic cloning efficiency in cattle.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit