Merck
  • Biomarker report from the phase II lamotrigine trial in secondary progressive MS - neurofilament as a surrogate of disease progression.
Biomarker report from the phase II lamotrigine trial in secondary progressive MS - neurofilament as a surrogate of disease progression.
PloS one (2013-08-13)
Sharmilee Gnanapavan, Donna Grant, Steve Morant, Julian Furby, Tom Hayton, Charlotte E Teunissen, Valerio Leoni, Monica Marta, Robert Brenner, Jacqueline Palace, David H Miller, Raj Kapoor, Gavin Giovannoni
ABSTRACT
Lamotrigine trial in SPMS was a randomised control trial to assess whether partial blockade of sodium channels has a neuroprotective effect. The current study was an additional study to investigate the value of neurofilament (NfH) and other biomarkers in predicting prognosis and/or response to treatment. SPMS patients who attended the NHNN or the Royal Free Hospital, UK, eligible for inclusion were invited to participate in the biomarker study. Primary outcome was whether lamotrigine would significantly reduce detectable serum NfH at 0-12, 12-24 and 0-24 months compared to placebo. Other serum/plasma and CSF biomarkers were also explored. Treatment effect by comparing absolute changes in NfH between the lamotrigine and placebo group showed no difference, however based on serum lamotrigine adherence there was significant decline in NfH (NfH 12-24 months p=0.043, Nfh 0-24 months p=0.023). Serum NfH correlated with disability: walking times, 9-HPT (non-dominant hand), PASAT, z-score, MSIS-29 (psychological) and EDSS and MRI cerebral atrophy and MTR. Other biomarkers explored in this study were not found to be significantly associated, aside from that of plasma osteopontin. The relations between NfH and clinical scores of disability and MRI measures of atrophy and disease burden support NfH being a potential surrogate endpoint complementing MRI in neuroprotective trials and sample sizes for such trials are presented here. We did not observe a reduction in NfH levels between the Lamotrigine and placebo arms, however, the reduction in serum NfH levels based on lamotrigine adherence points to a possible neuroprotective effect of lamotrigine on axonal degeneration.
Sigma-Aldrich
Glial Fibrillary Acidic Protein, Human Brain
Sigma-Aldrich
ChemiKine Brain Derived Neurotrophic Factor, Sandwich ELISA, This ChemiKine Brain Derived Neurotrophic Factor, Sandwich ELISA is used to measure & quantify BDNF levels in Neuroscience research.
Sigma-Aldrich
Phosphorylated Neurofilament (pNF-H) Sandwich ELISA Kit, The Phosphorylated Neurofilament H (pNF-H) kit is a sandwich enzyme immunoassay (EIA), which measures pNF-H.