Merck
  • 3D QSAR of novel estrogen-RGD peptide conjugates: getting insight into structural dependence of anti-osteoporosis activity and side effect of estrogen in ERT.
3D QSAR of novel estrogen-RGD peptide conjugates: getting insight into structural dependence of anti-osteoporosis activity and side effect of estrogen in ERT.
Bioorganic & medicinal chemistry (2009-04-24)
Ming Zhao, Jiangyuan Liu, Xiaoyi Zhang, Li Peng, Chunyu Li, Shiqi Peng
ABSTRACT
To explore the structural dependence of the oral potency and side effect of estrogen-RGD peptide conjugates, here six novel conjugates were prepared via introducing RGD-tetrapeptides into both 3- and 17-positions of estradiol, and introducing RGD-octapeptides into 3-position of estrone. In an ovariectomized mouse model they exhibited higher anti-osteoporosis activity and lower side effect than estrogen. For 3D QSAR analysis the anti-osteoporosis activities of nine known conjugates estrogen-RGD tetrapeptide conjugates were also provided. Using Cerius(2) module their 3D QSAR analysis was performed, four equations with high r(2) values were established, and the structural dependence of the oral potency and side effect of them was elucidated.
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture