Merck
  • Condensed bridgehead nitrogen heterocyclic system: synthesis and pharmacological activities of 1,2,4-triazolo-[3,4-b]-1,3,4-thiadiazole derivatives of ibuprofen and biphenyl-4-yloxy acetic acid.
Condensed bridgehead nitrogen heterocyclic system: synthesis and pharmacological activities of 1,2,4-triazolo-[3,4-b]-1,3,4-thiadiazole derivatives of ibuprofen and biphenyl-4-yloxy acetic acid.
European journal of medicinal chemistry (2007-11-21)
Mohd Amir, Harish Kumar, S A Javed
ABSTRACT
Several 3,6-disubstituted-1,2,4-triazolo-[3,4-b]-1,3,4-thiadiazoles were prepared by condensation of 4-amino-5-substituted-3-mercapto-(4H)-1,2,4-triazoles (3a,b) with various substituted aromatic acids and aryl/alkyl isothiocyanates through a one-pot reaction. These compounds were investigated for their anti-inflammatory, analgesic, ulcerogenic, lipid peroxidation, antibacterial and antifungal activities. Some of the synthesized compounds showed potent anti-inflammatory activity along with minimal ulcerogenic effect and lipid peroxidation, compared to those of ibuprofen and flurbiprofen. Some of the tested compounds also showed moderate antimicrobial activity against tested bacterial and fungal strains.
Product Description
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Diphenylacetic acid, 99%
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Flurbiprofen, cyclooxygenase inhibitor