Merck
  • Salinomycin-induced polyneuropathy in cats: morphologic and epidemiologic data.
Salinomycin-induced polyneuropathy in cats: morphologic and epidemiologic data.
Veterinary pathology (1999-03-31)
J S van der Linde-Sipman, T S van den Ingh, J J van nes, H Verhagen, J G Kersten, A C Beynen, R Plekkringa
ABSTRACT
In April 1996, an outbreak of toxic polyneuropathy in cats occurred in the Netherlands. All cats had been fed one of two brands of dry cat food from one manufacturer. Chemical analyses of these foods, stomach contents, and liver and kidney of affected cats revealed contamination with the ionophor salinomycin. Epidemiologic and clinical data were collected from 823 cats, or about 1% of the cats at risk. In 21 affected cats, postmortem examination was performed. The affected cats had acute onset of lameness and paralysis of the hindlimbs followed by the forelimbs. Clinical and pathologic examination indicated a distal polyneuropathy involving both the sensory and motor nerves.
Sigma-Aldrich
Salinomycin, from Streptomyces albus, ≥98% (HPLC)