- Prognostic and Monitoring Value of Circulating Tumor Cells in Adrenocortical Carcinoma: A Preliminary Monocentric Study.
Prognostic and Monitoring Value of Circulating Tumor Cells in Adrenocortical Carcinoma: A Preliminary Monocentric Study.
Adrenocortical carcinoma (ACC), a rare and aggressive neoplasia, presents poor prognosis when metastatic at diagnosis and limited therapies are available. Specific and sensitive markers for early diagnosis and a monitoring system of therapy and tumor evolution are urgently needed. The liquid biopsy represents a source of tumor material within a minimally invasive blood draw that allows the recovery of circulating tumor cells (CTCs). CTCs have been recently shown to be detectable in ACC. In the present paper, we evaluated the prognostic value of CTCs obtained by size-filtration in a small pilot cohort of 19 ACC patients. We found CTCs in 68% of pre-surgery and in 38% of post-surgery blood samples. In addition, CTC clusters (CTMs) and cancer associated macrophages (CAMLs) were detectable in some ACC patients. The median number of CTCs significantly decreased after the mass removal. Finally, stratifying patients in high and low pre-surgery CTC number groups, assuming the 75th percentile CTC value as cut-off, CTCs significantly predicted patients' overall survival (log rank = 0.005), also in a multivariate analysis adjusted for age and tumor stage. In conclusion, though preliminary and performed in a small cohort of patients, our study suggests that CTC number may represent a promising marker for prognosis and disease monitoring in ACC.