Merck
  • Doxorubicin-induced oxidative stress differentially regulates proteolytic signaling in cardiac and skeletal muscle.
Doxorubicin-induced oxidative stress differentially regulates proteolytic signaling in cardiac and skeletal muscle.
American journal of physiology. Regulatory, integrative and comparative physiology (2019-11-28)
Ryan N Montalvo, Vivian Doerr, Kisuk Min, Hazel H Szeto, Ashley J Smuder
ABSTRACT
Doxorubicin (DOX) is a highly effective antineoplastic agent used in cancer treatment. Unfortunately, clinical use of DOX is limited due to the development of dose-dependent toxicity to cardiac and respiratory (i.e., diaphragm) muscles. After administration, DOX preferentially localizes to the inner mitochondrial membrane, where it promotes cellular toxicity via enhanced mitochondrial reactive oxygen species (ROS) production. Although recent evidence suggests that amelioration of mitochondrial ROS emission preserves cardiorespiratory muscle function following DOX treatment, the mechanisms responsible for this protection remain unknown. Therefore, we tested the hypothesis that DOX-induced mitochondrial ROS production is required to stimulate pathological signaling by the autophagy/lysosomal system (ALS), the ubiquitin-proteasome pathway (UPP), and the unfolded protein response (UPR). Cause and effect were determined by administration of the mitochondria-targeted peptide SS-31 to DOX-treated animals. Interestingly, while SS-31 abrogated aberrant ROS emission in cardiorespiratory muscles of DOX-treated animals, our results revealed muscle-specific regulation of effector pathways. In the heart, SS-31 prevented DOX-induced proteolytic signaling through the ALS and UPP. In contrast, ALS signaling was inhibited by SS-31 in the diaphragm, but the UPP was not affected. UPR signaling was activated in both muscles at eukaryotic translation initiation factor 2α (eIF2α) S51 in the heart and diaphragm of DOX-treated animals and was attenuated with SS-31 treatment in both tissues. However, downstream signaling of eIF2α (activating transcription factor 4 and CCAAT/enhancer-binding protein homologous protein) was diminished in the heart but upregulated in the diaphragm with DOX. Collectively, these results show that DOX-induced ROS production plays distinct roles in the regulation of cardiac and diaphragm muscle proteolysis.
Sigma-Aldrich
Anti-phospho-ULK1 (Ser555) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution