Merck
  • Efficient COI barcoding using high throughput single-end 400 bp sequencing.
Efficient COI barcoding using high throughput single-end 400 bp sequencing.
BMC genomics (2020-12-06)
Chentao Yang, Yuxuan Zheng, Shangjin Tan, Guanliang Meng, Wei Rao, Caiqing Yang, David G Bourne, Paul A O'Brien, Junqiang Xu, Sha Liao, Ao Chen, Xiaowei Chen, Xinrui Jia, Ai-Bing Zhang, Shanlin Liu
ABSTRACT
Over the last decade, the rapid development of high-throughput sequencing platforms has accelerated species description and assisted morphological classification through DNA barcoding. However, the current high-throughput DNA barcoding methods cannot obtain full-length barcode sequences due to read length limitations (e.g. a maximum read length of 300 bp for the Illumina's MiSeq system), or are hindered by a relatively high cost or low sequencing output (e.g. a maximum number of eight million reads per cell for the PacBio's SEQUEL II system). Pooled cytochrome c oxidase subunit I (COI) barcodes from individual specimens were sequenced on the MGISEQ-2000 platform using the single-end 400 bp (SE400) module. We present a bioinformatic pipeline, HIFI-SE, that takes reads generated from the 5' and 3' ends of the COI barcode region and assembles them into full-length barcodes. HIFI-SE is written in Python and includes four function modules of filter, assign, assembly and taxonomy. We applied the HIFI-SE to a set of 845 samples (30 marine invertebrates, 815 insects) and delivered a total of 747 fully assembled COI barcodes as well as 70 Wolbachia and fungi symbionts. Compared to their corresponding Sanger sequences (72 sequences available), nearly all samples (71/72) were correctly and accurately assembled, including 46 samples that had a similarity score of 100% and 25 of ca. 99%. The HIFI-SE pipeline represents an efficient way to produce standard full-length barcodes, while the reasonable cost and high sensitivity of our method can contribute considerably more DNA barcodes under the same budget. Our method thereby advances DNA-based species identification from diverse ecosystems and increases the number of relevant applications.
Sigma-Aldrich
Sodium chloride solution, technical, ~26% (saturated in water at 20°C, AT)