Merck
  • The scaffolding protein calpain-3 has multiple distributions in embryonic chick muscle cells and it is essential for the formation of muscle fibers.
The scaffolding protein calpain-3 has multiple distributions in embryonic chick muscle cells and it is essential for the formation of muscle fibers.
Tissue & cell (2020-09-16)
Ivone de Andrade Rosa, Stephany Corrêa, Manoel Luis Costa, Claudia Mermelstein
ABSTRACT
CAPN3 is a muscle-specific and an intrinsically disordered protein. Thus, as a scaffolding protein CAPN3 could play a role during early stages of myogenesis. To test this hypothesis, we studied the distribution and function of CAPN3 during myogenesis using embryonic chick muscle cells grown in vitro. Super-resolution microscopy showed CAPN3 distribution in (i) amorphous patches in myoblasts, (ii) a region near the nuclei of myotubes; (iii) adhesion plaques in myotubes, (iv) stress fiber-like structures in myotubes, and (v) filaments in fibroblasts. Downregulation of CAPN3 induced a decrease in the number of muscle cells and in the size of myotubes formed. These data show a diverse intracellular distribution of CAPN3, compatible with a scaffolding protein, and suggest a multitude of different interactions of CAPN3 with other partners during muscle formation.
Sigma-Aldrich
MISSION® esiRNA, targeting human CAPN3, RP11-164J13.1 (1)