Merck
  • Delivering Cas9/sgRNA ribonucleoprotein (RNP) by lentiviral capsid-based bionanoparticles for efficient 'hit-and-run' genome editing.
Delivering Cas9/sgRNA ribonucleoprotein (RNP) by lentiviral capsid-based bionanoparticles for efficient 'hit-and-run' genome editing.
Nucleic acids research (2019-07-13)
Pin Lyu, Parisa Javidi-Parsijani, Anthony Atala, Baisong Lu
ABSTRACT
Transient expression of the CRISPR/Cas9 machinery will not only reduce risks of mutagenesis from off-target activities, but also decrease possible immune response to Cas9 protein. Building on our recent developing of a system able to package up to 100 copies of Cas9 mRNA in each lentivirus-like particle (LVLP) via the specific interaction between aptamer and aptamer-binding proteins (ABP), here we develop a lentiviral capsid-based bionanoparticle system, which allows efficient packaging of Cas9/sgRNA ribonucleoprotein (RNP). We show that replacing the Tetraloop of sgRNA scaffold with a com aptamer preserves the functions of the guide RNA, and the com-modified sgRNA can package Cas9/sgRNA RNP into lentivirus-like particles via the specific interactions between ABP and aptamer, and sgRNA and Cas9 protein. These RNP bionanoparticles generated Indels on different targets in different cells with efficiencies similar to or better than our recently described Cas9 mRNA LVLPs. The new system showed fast action and reduced off-target rates, and makes it more convenient and efficient in delivering Cas9 RNPs for transient Cas9 expression and efficient genome editing.
Sigma-Aldrich
Anti-Cas9 Antibody, clone 6F7, clone 6F7, from mouse