Merck
  • Study of Moisture-Curable Hybrid NIPUs Based on Glycerol with Various Diamines: Emergent Advantages of PDMS Diamines.
Study of Moisture-Curable Hybrid NIPUs Based on Glycerol with Various Diamines: Emergent Advantages of PDMS Diamines.
ACS omega (2020-12-08)
Georges R Younes, Gareth Price, Yves Dandurand, Milan Maric
ABSTRACT
A sol/gel curing method is used in this work to synthesize hybrid partially bio-based polyhydroxyurethanes (PHUs) from dicarbonates derived from glycerol and various diamines. The method consists of end-capping the PHU prepolymers with moisture-sensitive groups, so sealants and adhesives can be produced from partially sustainable hybrid PHUs (HPHUs), similar to their preparation from end-capped conventional polyurethanes. Diglycerol dicarbonate (DGC) is synthesized and polymerized with different diamines of various chain lengths, and the resulting structural and thermal properties of the PHUs are qualitatively and quantitively characterized. This characterization led to two potential candidates: PHU 4, made of DGC and a poly(propylene glycol) diamine, and PHU 10, prepared from DGC and a poly(dimethylsiloxane) diamine. These polymers, with respective relative number-average molecular weights of 3200 and 7400 g/mol, are end-capped and left to cure under ambient lab conditions (22 °C and 40-50% humidity), and the curing processes are monitored rheologically. Notably, moisture curing does not require any catalyst. The chemical stability of the resulting hybrid PHUs (HPHUs) 4 and 10 in pure water is investigated to check the viability of applying them under outdoor conditions. Only HPHU 10 is found to be resistant to water and shows hydrophobicity with a contact angle of 109°. Tensile tests are conducted on HPHU 10 samples cured under lab conditions for a week and others cured for another week while being immersed in water. The mechanical properties, tensile strength and elongation at break, improve with the samples cured in water, indicating the high-water repellency of HPHU 10.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Triglycerol, technical
Sigma-Aldrich
1,10-Diaminodecane, 97%