Merck
  • Synthesis, aerobic cytotoxicity, and radiosensitizing activity of novel 2,4-dinitrophenylamine tethered 5-fluorouracil and hydroxyurea.
Synthesis, aerobic cytotoxicity, and radiosensitizing activity of novel 2,4-dinitrophenylamine tethered 5-fluorouracil and hydroxyurea.
Bioorganic & medicinal chemistry letters (2006-09-23)
Ali Khalaj, Ali Reza Doroudi, Seyed Nasser Ostad, Mohammad Reza Khoshayand, Mohammad Babai, Neda Adibpour
ABSTRACT
Two novel dual functional agents, 3[3-(2,4-dinitro-phenylamino)-propyl]-5-fluoro-1H-pyrimidine-2,4-dione 7 and N-[3-(2,4-dinitro-phenylamino)-propoxy]urea 8, resulting from linkage of 2,4-dinitrophenylamine through three carbon atoms with 5-fluorouracil 5 and hydroxyurea 6, respectively, were prepared and their in vitro aerobic cytotoxicities in HT-29 cell line with and without radiation were determined. Compounds 7 and 8 unlike their components were not cytotoxic but showed radiosensitizing activity.
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications