Merck
  • Targeting PKCδ as a Therapeutic Strategy against Heterogeneous Mechanisms of EGFR Inhibitor Resistance in EGFR-Mutant Lung Cancer.
Targeting PKCδ as a Therapeutic Strategy against Heterogeneous Mechanisms of EGFR Inhibitor Resistance in EGFR-Mutant Lung Cancer.
Cancer cell (2018-12-12)
Pei-Chih Lee, Yueh-Fu Fang, Hirohito Yamaguchi, Wei-Jan Wang, Tse-Ching Chen, Xuan Hong, Baozhen Ke, Weiya Xia, Yongkun Wei, Zhengyu Zha, Yan Wang, Han-Pin Kuo, Chih-Wei Wang, Chih-Yen Tu, Chia-Hung Chen, Wei-Chien Huang, Shu-Fen Chiang, Lei Nie, Junwei Hou, Chun-Te Chen, Longfei Huo, Wen-Hao Yang, Rong Deng, Katsuya Nakai, Yi-Hsin Hsu, Shih-Shin Chang, Tai-Jan Chiu, Jun Tang, Ran Zhang, Li Wang, Bingliang Fang, Ting Chen, Kwok-Kin Wong, Jennifer L Hsu, Mien-Chie Hung
ABSTRACT
Multiple mechanisms of resistance to epidermal growth factor receptor (EGFR) tyrosine kinase inhibitors (TKIs) have been identified in EGFR-mutant non-small cell lung cancer (NSCLC); however, recurrent resistance to EGFR TKIs due to the heterogeneous mechanisms underlying resistance within a single patient remains a major challenge in the clinic. Here, we report a role of nuclear protein kinase Cδ (PKCδ) as a common axis across multiple known TKI-resistance mechanisms. Specifically, we demonstrate that TKI-inactivated EGFR dimerizes with other membrane receptors implicated in TKI resistance to promote PKCδ nuclear translocation. Moreover, the level of nuclear PKCδ is associated with TKI response in patients. The combined inhibition of PKCδ and EGFR induces marked regression of resistant NSCLC tumors with EGFR mutations.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-PLCG1 (pTyr783) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Dvl-PDZ Domain Inhibitor II, The Dvl-PDZ Domain Inhibitor II, also referenced under CAS 294891-81-9, controls the biological activity of Dvl-PDZ. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
KN-62, A cell-permeable, reversible, and selective inhibitor of CaM kinase II (Ki = 900 nM for rat brain CaM kinase II) that binds directly to the calmodulin binding site of the enzyme.