Merck
  • Resveratrol-primed exosomes strongly promote the recovery of motor function in SCI rats by activating autophagy and inhibiting apoptosis via the PI3K signaling pathway.
Resveratrol-primed exosomes strongly promote the recovery of motor function in SCI rats by activating autophagy and inhibiting apoptosis via the PI3K signaling pathway.
Neuroscience letters (2020-07-20)
Yue Fan, Yuanlong Li, Silin Huang, Huali Xu, Hui Li, Beixing Liu
ABSTRACT
Spinal cord injury (SCI) is a traumatic condition of the central nervous system (CNS) that can cause paralysis of the limbs. The molecular mechanisms of neural repair following SCI remain unclear and no effective treatment for SCI currently exists, since drugs have difficulty crossing the blood-brain barrier (BBB). The present study aimed to investigate whether exosomes could be used as specific carriers of resveratrol for induction of neuronal autophagy both in vitro and in vivo for the treatment of SCI. The results indicate that exosomes are able to enhance the solubility of resveratrol and enhance penetration of the drug through the BBB, thereby increasing its concentration in the CNS. Exosomes derived from resveratrol-treated primary microglia (Exo + Res) assisted the rehabilitation of paralyzed limbs in rats. Restoration of neural function following SCI was mediated through increased induction of autophagy and inhibition of apoptosis of neurons both in vitro and in vivo via activation of the PI3K signaling pathway. The mechanism of action of Exo + Res may be associated with the PI3K inhibitor 3-methyladenine (3-MA) in primary spinal neurons. The results suggest that Exo + Res are highly effective at crossing the BBB with good stability, suggesting they have potential for enhancing targeted drug delivery and the recovery of neuronal function in SCI therapy, likely associated with the induction of autophagy and inhibition of apoptosis via the PI3K signaling pathway.
Sigma-Aldrich
Tofisopam, ≥98% (HPLC), solid