Merck
  • Hepatic STAMP2 mediates recombinant FGF21-induced improvement of hepatic iron overload in nonalcoholic fatty liver disease.
Hepatic STAMP2 mediates recombinant FGF21-induced improvement of hepatic iron overload in nonalcoholic fatty liver disease.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-07-29)
Hye Young Kim, Woo Young Kwon, Joon Beom Park, Mi Hwa Lee, Yoo Jin Oh, SungHwan Suh, Yang Hyun Baek, Jin Sook Jeong, Young Hyun Yoo
ABSTRACT
Although previous studies have shown that the administration of fibroblast growth factor 21 (FGF21) reverses hepatic steatosis, the mechanism by which FGF21 exerts a therapeutic effect on nonalcoholic fatty liver disease (NAFLD) is not yet entirely understood. We previously demonstrated that hepatic six transmembrane protein of prostate 2 (STAMP2) may represent a suitable target for NAFLD. We investigated the mechanism underlying the therapeutic effect of recombinant FGF21 on NAFLD, focusing on the involvement of hepatic STAMP2. In this study, we used human nonalcoholic steatosis patient pathology samples, C57BL/6 mice for a high-fat diet (HFD)-induced in vivo NAFLD model, and used human primary hepatocytes and HepG2 cells for oleic acid (OA)-induced in vitro NAFLD model. We observed that recombinant FGF21 treatment ameliorated hepatic steatosis and insulin resistance through the upregulation of STAMP2 expression. We further observed hepatic iron overload (HIO) and reduced iron exporter, ferroportin expression in the liver samples obtained from human NAFLD patients, and HFD-induced NAFLD mice and in OA-treated HepG2 cells. Importantly, recombinant FGF21 improved HIO through the hepatic STAMP2-mediated upregulation of ferroportin expression. Our data suggest that hepatic STAMP2 may represent a suitable therapeutic intervention target for FGF21-induced improvement of NAFLD accompanying HIO.
Sigma-Aldrich
MISSION® esiRNA, targeting human STEAP4
Sigma-Aldrich
Ammonium iron(III) citrate, reagent grade, powder
Sigma-Aldrich
Ammonium citrate tribasic, ≥97% (titration)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)