Merck
Regulation of Retinal Melanopsin on Lens-induced Myopia in Guinea Pigs.
Optometry and vision science : official publication of the American Academy of Optometry (2020-07-23)
Wei Zheng, Yao Chen, Xuezhi Zhou, Xueyong Zhang, Yuanyuan Chen, Xinyu Guan, Junfeng Mao
ABSTRACT
Investigation of the mechanism and the role of melanopsin in lens-induced myopia is necessary to find out potential targets in the prevention of myopia development. We aimed to study the effect and mechanism of retinal melanopsin on lens-induced myopia in guinea pigs, as well as the interactions between melanopsin and other myopic regulation neurotransmitters such as dopamine and melatonin, and to explore the possible role of melanopsin in the prevention of myopia development. Twenty-day-old tricolor guinea pigs were randomly divided into four groups: control group, defocus group, defocus + AA92593 group, and defocus + dimethyl sulfoxide (DMSO) group. The defocus eyes wore -6.00 D lens. In the defocus + AA92593 group, the vitreous cavities were injected with melanopsin antagonist AA92593. In the defocus + DMSO group, the vitreous cavities were injected with 5% DMSO as the administration control. The expression of retinal melanopsin protein was measured with immunofluorescence staining and Western blot. The content of dopamine and melatonin in the retina was determined by the high-performance liquid chromatography electrochemical method. Compared with the defocus group, intravitreal injection of AA92593 resulted in increased axial length of the defocus eyes (defocus, 8.05 ± 0.09 mm; defocus + AA92593, 8.15 ± 0.11 mm; P = .008), lower refractive degree (defocus, -1.98 ± 0.82 D; defocus + AA92593, -2.59 ± 0.97 D; P = .05), decreased relative expression of retinal melanopsin protein (defocus, 0.67 ± 0.11; defocus + AA92593, 0.20 ± 0.06; P < .0001), and increased melatonin content in the defocus eyes (defocus, 0.38 ± 0.09 ng/mg; defocus + AA92593, 0.55 ± 0.13 ng/mg; P = .01), but it had no obvious effect on dopamine content (defocus, 0.64 ± 0.18 ng/mg; defocus + AA9259, 0.61 ± 0.17 ng/mg; P > .99). The melatonin content of retina in the defocus + AA92593 group was correlated with refractive error (Pearson correlation coefficient = -0.68, P = .006) and eye axis length (Pearson correlation coefficient = 0.74, P = .02). Retinal melanopsin has inhibitory effect on lens-induced myopia development in guinea pigs, and such effect may be related to retinal melatonin.
Sigma-Aldrich
AA92593, ≥98% (HPLC)
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Dopamine hydrochloride, British Pharmacopoeia (BP) Reference Standard