Merck
  • Metabolomics by Gas Chromatography-Mass Spectrometry: Combined Targeted and Untargeted Profiling.
Metabolomics by Gas Chromatography-Mass Spectrometry: Combined Targeted and Untargeted Profiling.
Current protocols in molecular biology (2016-04-03)
Oliver Fiehn
ABSTRACT
Gas chromatography-mass spectrometry (GC-MS)-based metabolomics is ideal for identifying and quantitating small-molecule metabolites (<650 Da), including small acids, alcohols, hydroxyl acids, amino acids, sugars, fatty acids, sterols, catecholamines, drugs, and toxins, often using chemical derivatization to make these compounds sufficiently volatile for gas chromatography. This unit shows how GC-MS-based metabolomics allows integration of targeted assays for absolute quantification of specific metabolites with untargeted metabolomics to discover novel compounds. Complemented by database annotations using large spectral libraries and validated standard operating procedures, GC-MS can identify and semiquantify over 200 compounds from human body fluids (e.g., plasma, urine, or stool) per study. Deconvolution software enables detection of more than 300 additional unidentified signals that can be annotated through accurate mass instruments with appropriate data processing workflows, similar to untargeted profiling using liquid chromatography-mass spectrometry. GC-MS is a mature technology that uses not only classic detectors (quadrupole) but also target mass spectrometers (triple quadrupole) and accurate mass instruments (quadrupole-time of flight). This unit covers sample preparation from mammalian samples, data acquisition, quality control, and data processing.
Sigma-Aldrich
Fatty Acid Methyl Esters Standard Mixture
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterated Amino Acid Standard Mixture, suitable for mass spectrometry (MS)
Pricing and availability is not currently available.