Merck
CD45: all is not yet crystal clear.
Immunology (2006-01-21)
Nick Holmes
ABSTRACT
CD45 has been recognized as an important player in regulating signalling in lymphocytes. However, compared with tyrosine kinases, phosphatases are still poorly understood in terms of the details of their specificity and regulation. Here, the recent progress in understanding the biology of the first recognized receptor tyrosine phosphatase, CD45, is reviewed.