Merck
  • Production of fructooligosaccharides by Bacillus subtilis natto CCT7712 and their antiproliferative potential.
Production of fructooligosaccharides by Bacillus subtilis natto CCT7712 and their antiproliferative potential.
Journal of applied microbiology (2020-01-01)
A Magri, M R Oliveira, C Baldo, C A Tischer, D Sartori, M S Mantovani, M A P C Celligoi
ABSTRACT
Fructooligosaccharides (FOSs) known for their health properties and β-(2→6)-levan-type FOSs have shown prebiotic and immunomodulatory activities that overcome those of commercial β-(2→1)-FOSs, but costs do not favour their use. Moreover, FOSs can reach the bloodstream through the diet, and little is known about their direct effect on cells. The aim of this work was to produce high-content FOSs by Bacillus subtilis natto CCT7712 in a bioreactor using commercial sucrose and to evaluate their antiproliferative effects in OVCAR-3 cells. FOS production reached 173·60 g l-1 , 0·2 vvm aeration and uncontrolled pH. Levan-type FOSs, composed of β-(2 → 6) links and mainly GF3 (6-nystose), were identified using RMN spectroscopy, FT-IR and ESI-MS. FOSs decreased the viability and proliferation of OVCAR-3 cells, and the effects were associated with an increased pro-inflammatory response by the induction of IL-8 and TNF-α, and the repression of ER-β genes. The metabolic profiles showed disruption of cellular homeostasis that can be associated with a decrease in proliferation. The high production of levan-type FOSs from B. subtilis natto CCT7712 in a bioreactor was achieved, and they showed antiproliferative potential in OVCAR-3 cells. FOS could be a good target for future therapeutic studies and commercial use.
Product Description
Supelco
Nystose, analytical standard