Merck
  • Antimycotic Activity of Ozonized Oil in Liposome Eye Drops against Candida spp.
Antimycotic Activity of Ozonized Oil in Liposome Eye Drops against Candida spp.
Translational vision science & technology (2020-08-29)
Giuseppe Celenza, Roberto Iorio, Salvatore Cracchiolo, Sabrina Petricca, Ciro Costagliola, Benedetta Cinque, Bernardetta Segatore, Gianfranco Amicosante, Pierangelo Bellio
ABSTRACT
This study aims to investigate the antifungal activity and mechanism of action of ozonized oil eye drops in liposomes (Ozodrop), commercialized as eye lubricant for the treatment of dry eye syndrome and eye inflammation. The activity was tested against four clinical Candida species: Calbicans,Cglabrata,Ckrusei, and Corthopsilosis. The antifungal activity of the eye drop solution was ascertained by microdilution method in accordance with EUCAST obtaining the minimum inhibitory concentration for Ozodrop. The mechanism of action was further investigated in Calbicans by measuring cell vitality, intracellular reactive oxygen species production, levels of cellular and mitochondrial (∆Ψm) membrane potential, and the extent of membrane lipid peroxidation. All Candida isolates were susceptible to Ozodrop with minimum inhibitory concentration values ranging from 0.195% (v/v) for Cglabrata to 6.25% (v/v) for Corthopsilosis. After 1 hour of exposure at the minimum inhibitory concentration value about 30% of cells were killed, reaching about 70% at the highest Ozodrop value. After Ozodrop exposure, Calbicans showed cell membrane depolarization, increased levels of lipid peroxidation, depolarized ∆Ψm, and increased reactive oxygen species generation. The significant increases in reactive oxygen species production cause the accumulation of reactive oxygen species-associated damages leading to progressive Candida cell dysfunction. The antifungal activity of Ozodrop was demonstrated at concentrations several times lower than the concentration that can be retrieved in ocular surface after its application. The antifungal activity of the eye drops Ozodrop would represent an interesting off-label indication for a product basically conceived as an eye lubricant.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
3,3′-Dipropylthiacarbocyanine iodide, 98%