Merck
Pd(II)-catalyzed enantioselective C-H activation of cyclopropanes.
Journal of the American Chemical Society (2011-11-09)
Masayuki Wasa, Keary M Engle, David W Lin, Eun Jeong Yoo, Jin-Quan Yu
ABSTRACT
Systematic ligand development has led to the identification of novel mono-N-protected amino acid ligands for Pd(II)-catalyzed enantioselective C-H activation of cyclopropanes. A diverse range of organoboron reagents can be used as coupling partners, and the reaction proceeds under mild conditions. These results provide a new retrosynthetic disconnection for the construction of enantioenriched cis-substituted cyclopropanecarboxylic acids.
Sigma-Aldrich
Yu-Wasa Auxiliary, 97%
Sigma-Aldrich
Cyclopropanecarboxylic acid, 95%