Merck
  • NF-Y and Sp1/Sp3 are involved in the transcriptional regulation of the peptidylarginine deiminase type III gene (PADI3) in human keratinocytes.
NF-Y and Sp1/Sp3 are involved in the transcriptional regulation of the peptidylarginine deiminase type III gene (PADI3) in human keratinocytes.
The Biochemical journal (2006-05-05)
Sijun Dong, Takuya Kanno, Ayako Yamaki, Toshio Kojima, Masakazu Shiraiwa, Akira Kawada, Marie-Claire Méchin, Stéphane Chavanas, Guy Serre, Michel Simon, Hidenari Takahara
ABSTRACT
Human peptidylarginine deiminase type III gene (PADI3) encodes a crucial post-translational modification enzyme that converts protein-bound arginine residues into citrulline residues. Its expression is restricted to a few cell types, including keratinocytes in the granular layer of the epidermis and in the inner root sheath of hair follicles. In these cells, the enzyme is involved in terminal processing of intermediate filament-binding proteins such as filaggrin and trichohyalin. To study the molecular mechanisms that control the expression of PADI3 in human keratinocytes at the transcriptional level, we characterized its promoter region using human keratinocytes transfected with variously deleted fragments of the 5'-upstream region of PADI3 coupled to the luciferase gene. We found that as few as 129 bp upstream from the transcription initiation site were sufficient to direct transcription of the reporter gene. Electrophoretic mobility-shift and chromatin immunoprecipitation assays revealed that NF-Y (nuclear factor Y) and Sp1/Sp3 (specificity protein 1/3) bind to this region in vitro and in vivo. Moreover, mutation of the Sp1- or NF-Y-binding motif markedly reduced PADI3 promoter activity. Furthermore, Sp1 or NF-YA (NF-Y subunit) small interfering RNAs effectively diminished PADI3 expression in keratinocytes cultured in both low- and high-calcium medium. These data indicate that PADI3 expression is driven by Sp1/Sp3 and NF-Y binding to the promoter region.
Sigma-Aldrich
PADI-3 human, recombinant, expressed in baculovirus infected Sf9 cells, ≥60% (SDS-PAGE)