Merck
  • Occurrence of Halogenated Pollutants in Domestic and Occupational Indoor Dust.
Occurrence of Halogenated Pollutants in Domestic and Occupational Indoor Dust.
International journal of environmental research and public health (2020-05-31)
Giulia Simonetti, Patrizia Di Filippo, Carmela Riccardi, Donatella Pomata, Elisa Sonego, Francesca Buiarelli
ABSTRACT
The occurrence of halogenated organic pollutants in indoor dust can be high due to the presence of textile, electronic devices, furniture, and building materials treated with these chemicals. In this explorative study, we focused on emerging organic pollutants, such as novel brominated flame retardants (nBFRs) and some perfluoroalkyl substances, together with legacy polychlorinated biphenyls (PCBs) and polybrominated diphenyl ethers (BDEs) in settled dust collected in houses and workplaces such as one office and two electrotechnical and mechanical workshops. The total contribution of the investigated pollutants was lower in house and in office dusts except for few nBFRs (such as bis (2-ethylhexyl)-3,4,5,6-tetrabromo-phthalate at a concentration of 464.5 ng/g in a house and hexachlorocyclopentadienyldibromocyclooctane at 40.4 ng/g in the office), whereas in electrotechnical and mechanical workshops a high incidence of PCBs, BDEs, and nBFRs occurred (for example, BDE 209 at a concentration of 2368.0 ng/g and tetrabromobisphenol A at 32,320.1 ng/g in electrotechnical and mechanical workshops). Estimated daily intakes were also calculated, showing that domestic and occupational environments can lead to a similar contribution in terms of human exposure. The higher exposure contribution was associated to nBFRs, whose EDIs were in the range of 3968.2-555,694.2 pg/kg bw/day. To provide a complete view about the indoor contamination, in this investigation, we also included polycyclic aromatic hydrocarbons (PAHs) and their oxygenated and nitrated derivatives. Definitely, dust collection represents a simple, fast, and cost-effective sampling and dust contamination level can be a useful indicator of environment healthiness. Besides, the presented method can be a smart tool to provide a time and money saving technique to characterize 99 pollutants thanks to a single sample treatment.
Sigma-Aldrich
2,4,4,6-Tetrabromo-2,5-cyclohexadienone, 90%