Merck
High-throughput fluorescence assay for membrane-protein interaction.
Journal of lipid research (2013-09-06)
Hyunjin Kim, Hamid Samareh Afsari, Wonhwa Cho
ABSTRACT
Membrane-protein interaction plays key roles in a wide variety of biological processes. Although various methods have been employed to measure membrane binding of soluble proteins, a robust high-throughput assay that is universally applicable to all proteins is lacking at present. Here we report a new fluorescence quenching assay utilizing enhanced green fluorescence protein (EGFP)-fusion proteins and a lipid containing a dark quencher, N-dimethylaminoazobenzenesulfonyl-phosphatidylethanolamine (dabsyl-PE). The EGFP fluorescence emission intensity showed a large decrease (i.e., >50%) when EGFP-fusion proteins bound the vesicles containing 5 mol% dabsyl-PE. This simple assay, which can be performed using either a cuvette-based spectrofluorometer or a fluorescence plate reader, allowed rapid, sensitive, and accurate determination of lipid specificity and affinity for various lipid binding domains, including two pleckstrin homology domains, an epsin N-terminal homology domain, and a phox homology domain. The assay can also be applied to high-throughput screening of small molecules that modulate membrane binding of proteins.