Merck
  • Lysophosphatidylcholine and lyso-PAF display PAF-like activity derived from contaminating phospholipids.
Lysophosphatidylcholine and lyso-PAF display PAF-like activity derived from contaminating phospholipids.
Journal of lipid research (2001-08-24)
G K Marathe, A R Silva, H C de Castro Faria Neto, L W Tjoelker, S M Prescott, G A Zimmerman, T M McIntyre
ABSTRACT
Lysophosphatidylcholine is an abundant component of plasma and oxidized LDL that displays several biological activities, some of which may occur through the platelet-activating factor (PAF) receptor. We find that commercial lysophosphatidylcholine, its alkyl homolog (lyso-PAF), and PAF all induce inflammation in a murine model of pleurisy. Hydrolysis of PAF to lyso-PAF by recombinant PAF acetylhydrolase abolished this eosinophilic infiltration, implying that lyso-PAF should not have displayed inflammatory activity. Saponification of lyso-PAF or PAF acetylhydrolase treatment of lyso-PAF or lysophosphatidylcholine abolished activity; neither lysolipid should contain susceptible sn-2 residues, suggesting contaminants account for the bioactivity. Lyso-PAF and to a lesser extent lysophosphatidylcholine stimulated Ca(2+) accumulation in 293 cells stably transfected with the human PAF receptor, and this was inhibited by specific PAF receptor antagonists. Again, treatment of lyso-PAF or lysophosphatidylcholine with recombinant PAF acetylhydrolase, a nonselective phospholipase A(2), or saponification of lyso-PAF destroyed the PAF-like activity, a result incompatible with lyso-PAF or lysophosphatidylcholine being the actual agonist. We conclude that neither lyso-PAF nor lysophosphatidylcholine is a PAF receptor agonist, nor are they inflammatory by themselves. We suggest that PAF or a PAF-like mimetic accounts for inflammatory effects of lysophosphatidylcholine and lyso-PAF.