- Fatty acid amide signaling molecules.
Fatty acid amide signaling molecules.
Bioorganic & medicinal chemistry letters (2010-09-08)
Cyrine Ezzili, Katerina Otrubova, Dale L Boger
PMID20817522
ABSTRACT
Key studies leading to the discovery and definition of the role of endogenous fatty acid amide signaling molecules are summarized.