Merck
  • Synthetic High-density Lipoprotein Nanodiscs for Personalized Immunotherapy Against Gliomas.
Synthetic High-density Lipoprotein Nanodiscs for Personalized Immunotherapy Against Gliomas.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-05-23)
Lindsay Scheetz, Padma Kadiyala, Xiaoqi Sun, Sejin Son, Alireza Hassani Najafabadi, Marisa Aikins, Pedro R Lowenstein, Anna Schwendeman, Maria G Castro, James J Moon
ABSTRACT
Gliomas are brain tumors with dismal prognoses. The standard-of-care treatments for gliomas include surgical resection, radiation, and temozolomide administration; however, they have been ineffective in providing significant increases in median survival. Antigen-specific cancer vaccines and immune checkpoint blockade may provide promising immunotherapeutic approaches for gliomas. We have developed immunotherapy delivery vehicles based on synthetic high-density lipoprotein (sHDL) loaded with CpG, a Toll-like receptor-9 agonist, and tumor-specific neoantigens to target gliomas and elicit immune-mediated tumor regression. We demonstrate that vaccination with neoantigen peptide-sHDL/CpG cocktail in combination with anti-PD-L1 immune checkpoint blocker elicits robust neoantigen-specific T-cell responses against GL261 cells and eliminated established orthotopic GL261 glioma in 33% of mice. Mice remained tumor free upon tumor cell rechallenge in the contralateral hemisphere, indicating the development of immunologic memory. Moreover, in a genetically engineered murine model of orthotopic mutant IDH1 (mIDH1) glioma, sHDL vaccination with mIDH1 neoantigen eliminated glioma in 30% of animals and significantly extended the animal survival, demonstrating the versatility of our approach in multiple glioma models. Overall, our strategy provides a general roadmap for combination immunotherapy against gliomas and other cancer types.