Merck
  • lncTUG1/miR-144-3p affect the radiosensitivity of esophageal squamous cell carcinoma by competitively regulating c-MET.
lncTUG1/miR-144-3p affect the radiosensitivity of esophageal squamous cell carcinoma by competitively regulating c-MET.
Journal of experimental & clinical cancer research : CR (2020-01-11)
Pan Wang, Zhuanbo Yang, Ting Ye, Fei Shao, Jiagen Li, Nan Sun, Jie He
ABSTRACT
Long noncoding RNAs (lncRNAs) are involved in the progression of various cancers and affect the response to radiotherapy. This study focused on clarifying the underlying mechanism by which lncTUG1 affects the radiosensitivity of esophageal squamous cell carcinoma (ESCC). lncTUG1, miR-144-3p and MET expression levels were detected in ESCC tissues and cells by qRT-PCR. Western blotting was used to examine the protein levels of MET, p-AKT and EGFR. The dual-luciferase reporter system and RNA immunoprecipitation (RIP) assays were used to confirm the interaction between lncTUG1 and miR-144-3p or miR-144-3p and MET. MTT, colony formation and flow cytometry assays were applied to examine the behavioral changes in EC9706 and KYSE30 cells. lncTUG1 was upregulated in ESCC cells and tissues, and lncTUG1 expression was associated with an advanced pathological stage. The bioinformatics analysis revealed that lncTUG1 could specifically bind to miR-144-3p, which was downregulated in ESCC. There was a negative correlation between lncTUG1 and miR-144-3p. LncTUG1 inhibition retarded proliferation and colony formation and induced apoptosis in ESCC cells. Moreover, lncTUG1 knockdown dramatically improved the effect of radiotherapy on ESCC development both in vivo and in vitro. Furthermore, MET was revealed as a downstream target of miR-144-3p and is downregulated by it. LncTUG1 promoted the progression of ESCC and elevated radiotherapy resistance in ESCC cells, accompanied by a high level of MET expression. Moreover, we found that knockdown of lncTUG1 enhanced the radiosensitivity of ESCC cells via the p-AKT signaling pathway. Our results indicate that lncTUG1 enhances the radiotherapy resistance of ESCC by lowering the miR-144-3p level and modulating the MET/EGFR/AKT axis.
Sigma-Aldrich
MISSION® esiRNA, targeting human TUG1