Merck
  • DOCK8 functions as an adaptor that links TLR-MyD88 signaling to B cell activation.
DOCK8 functions as an adaptor that links TLR-MyD88 signaling to B cell activation.
Nature immunology (2012-05-15)
Haifa H Jabara, Douglas R McDonald, Erin Janssen, Michel J Massaad, Narayanaswamy Ramesh, Arturo Borzutzky, Ingrid Rauter, Halli Benson, Lynda Schneider, Sachin Baxi, Mike Recher, Luigi D Notarangelo, Rima Wakim, Ghassan Dbaibo, Majed Dasouki, Waleed Al-Herz, Isil Barlan, Safa Baris, Necil Kutukculer, Hans D Ochs, Alessandro Plebani, Maria Kanariou, Gerard Lefranc, Ismail Reisli, Katherine A Fitzgerald, Douglas Golenbock, John Manis, Sevgi Keles, Reuben Ceja, Talal A Chatila, Raif S Geha
ABSTRACT
The adaptors DOCK8 and MyD88 have been linked to serological memory. Here we report that DOCK8-deficient patients had impaired antibody responses and considerably fewer CD27(+) memory B cells. B cell proliferation and immunoglobulin production driven by Toll-like receptor 9 (TLR9) were considerably lower in DOCK8-deficient B cells, but those driven by the costimulatory molecule CD40 were not. In contrast, TLR9-driven expression of AICDA (which encodes the cytidine deaminase AID), the immunoglobulin receptor CD23 and the costimulatory molecule CD86 and activation of the transcription factor NF-κB, the kinase p38 and the GTPase Rac1 were intact. DOCK8 associated constitutively with MyD88 and the tyrosine kinase Pyk2 in normal B cells. After ligation of TLR9, DOCK8 became tyrosine-phosphorylated by Pyk2, bound the Src-family kinase Lyn and linked TLR9 to a Src-kinase Syk-transcription factor STAT3 cascade essential for TLR9-driven B cell proliferation and differentiation. Thus, DOCK8 functions as an adaptor in a TLR9-MyD88 signaling pathway in B cells.
Sigma-Aldrich
Anti-Src Antibody, CT, clone NL19 | 04-772, clone NL19, Upstate®, from rabbit
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 9E10, clone 9E10, Upstate®, from mouse
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse