Merck
  • Practical radical cyclizations with arylboronic acids and trifluoroborates.
Practical radical cyclizations with arylboronic acids and trifluoroborates.
Organic letters (2011-09-20)
Jonathan W Lockner, Darryl D Dixon, Rune Risgaard, Phil S Baran
ABSTRACT
Practical radical cyclizations using organoboronic acids and trifluoroborates take place in water, open to air, and in a scalable fashion employing catalytic silver nitrate and stoichiometric potassium persulfate. Both Pschorr-type cyclizations and tandem radical cyclization/trap cascades are described, illustrating the utility of these mild conditions for the generation of polycyclic scaffolds.