Merck
  • Interaction of fibroblasts and induced pluripotent stem cells with poly(vinyl alcohol)-based hydrogel substrates.
Interaction of fibroblasts and induced pluripotent stem cells with poly(vinyl alcohol)-based hydrogel substrates.
Journal of biomedical materials research. Part B, Applied biomaterials (2019-06-30)
Eduardo de O Sanguinet, Nataly M Siqueira, Felipe de C Menezes, Gisele M Rasia, Nívia Lothhammer, Rosane M D Soares, Flávio V Meirelles, Fabiana F Bressan, Adriana Bos-Mikich
ABSTRACT
Induced pluripotent stem cells (iPSCs) provide a promising means of creating custom-tailored cell lines for cellular therapies. Their application in regenerative medicine, however, depends on the possibility that the maintenance and differentiation of cells and organs occur under defined conditions. One major component of stem cell culture systems is the substrate, where the cells must attach and proliferate. The present study aimed to investigate the putative cytotoxic effects of poly(vinyl alcohol) (PVA)-based matrices on the in vitro culture of mouse fetal fibroblasts. In addition, the PVA-based hydrogels were used to determine the capacity of bovine induced pluripotent stem cells (biPSCs) to adhere and proliferate on synthetic substrates. Our results show that both cell types interacted with the substrate and presented proliferation during culture. The biPSCs formed new colonies when cell suspensions were placed onto the hydrogel surface for culture. These results may represent a new characterized xeno-free clinical grade culture system to be widely applied in cell-based therapies.
Sigma-Aldrich
1,2,3,4-Butanetetracarboxylic acid, 99%